siRNA Details
| Gene Symbol: Eml6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146016.2 | NP_666128.2 | 37 | 5980 | ||
| GenBank mRNA | AK081418.1 | - | 2 | 331 | ||
| BC024726.1 | - | 9 | 1415 | 5362808 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eml6| Gene Symbol: Eml6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146016.2 | NP_666128.2 | 37 | 5980 | ||
| GenBank mRNA | AK081418.1 | - | 2 | 331 | ||
| BC024726.1 | - | 9 | 1415 | 5362808 | ||