siRNA Details
| Gene Symbol: Rtn4rl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177708.5 | NP_808376.1 | 2 | 1457 | ||
| GenBank mRNA | AK033286.1 | - | 2 | 1455 | ||
| AK140456.1 | - | 2 | 1458 | |||
| AY250218.1 | - | 1 | 1168 | |||
| BC030471.1 | - | 2 | 1661 | 5368560 | ||

Rtn4rl1
Genomic Map