siRNA Details
| Gene Symbol: Slfn10-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041394.1 | - | 4 | 1563 | ||
| AK144698.1 | - | 5 | 2156 | |||
| AK220241.1 | - | 5 | 2280 | |||
| AY261802.1 | - | 5 | 2155 | |||
| AY261803.1 | - | 5 | 2124 | |||
| BC115828.1 | - | 1047 | ||||

Slfn10-ps
Genomic Map