siRNA Details
| Gene Symbol: Usp32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029934.1 | NP_001025105.1 | 25, 26 | 3222 | ||
| GenBank mRNA | AK148221.1 | - | 25, 26 | 3222 | ||
| BC040268.1 | - | 2, 3 | 187 | |||
| BC051399.1 | - | 2, 3 | 260 | |||
| BC058994.1 | - | 6, 7 | 763 | |||

Usp32
Genomic Map