siRNA Details
Gene Symbol: Usp32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029934.1 | NP_001025105.1 | 34 | 4860 | ||
GenBank mRNA | AK032362.1 | - | 3 | 157 | ||
AK140739.1 | - | 3 | 233 | |||
BC040268.1 | - | 11 | 1825 | |||
BC051399.1 | - | 11 | 1898 | |||
BC058994.1 | - | 15 | 2401 |
