siRNA Details
Gene Symbol: Usp32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029934.1 | NP_001025105.1 | 32 | 4694 | ||
GenBank mRNA | AK013783.1 | - | 1 | 885 | ||
AK140739.1 | - | 1 | 67 | |||
BC040268.1 | - | 9 | 1659 | |||
BC051399.1 | - | 9 | 1732 | |||
BC058994.1 | - | 13 | 2235 |
