siRNA Details
| Gene Symbol: Coro1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011778.2 | NP_035908.1 | 8 | 949 | ||
| GenBank mRNA | AF143956.1 | - | 8 | 956 | ||
| AK008947.1 | - | 4 | 389 | |||
| AK149639.1 | - | 8 | 959 | |||
| AK159329.1 | - | 6 | 643 | |||
| AK197317.1 | - | 3 | 210 | |||
| AK205268.1 | - | 3 | 153 | |||
| BC029326.1 | - | 8 | 908 | |||
| BC131944.1 | - | 8 | 896 | 40130221 | ||
| BC132617.1 | - | 8 | 896 | 40130894 | ||

Coro1b
Genomic Map