siRNA Details
| Gene Symbol: Coro1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011778.2 | NP_035908.1 | 10 | 1203 | ||
| GenBank mRNA | AF143956.1 | - | 10 | 1210 | ||
| AK008947.1 | - | 6 | 643 | |||
| AK149639.1 | - | 10 | 1213 | |||
| AK159329.1 | - | 8 | 895 | |||
| AK184918.1 | - | 3 | 251 | |||
| AK215642.1 | - | 3 | 251 | |||
| BC029326.1 | - | 10 | 1162 | |||
| BC131944.1 | - | 10 | 1150 | 40130221 | ||
| BC132617.1 | - | 10 | 1150 | 40130894 | ||

Coro1b
Genomic Map