siRNA Details
| Gene Symbol: Gpatch8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159492.1 | NP_001152964.1 | 8 | 2828 | ||
| GenBank mRNA | AK122307.1 | - | 1 | 278 | ||
| AK142570.1 | - | 1 | 72 | |||
| AK156405.1 | - | 1 | 426 | |||
| BC035255.1 | - | 6 | 2887 | |||

Gpatch8
Genomic Map