siRNA Details
| Gene Symbol: Otop2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172801.2 | NP_766389.2 | 7, 8 | 1852 | ||
| GenBank mRNA | AB087504.1 | - | 6, 7 | 1737 | ||
| AK028866.1 | - | 7, 8 | 1853 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Otop2| Gene Symbol: Otop2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172801.2 | NP_766389.2 | 7, 8 | 1852 | ||
| GenBank mRNA | AB087504.1 | - | 6, 7 | 1737 | ||
| AK028866.1 | - | 7, 8 | 1853 | |||