siRNA Details
| Gene Symbol: Cdr2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080929.1 | NP_001074398.1 | 2 | 561 | ||
| GenBank mRNA | BC144823.1 | - | 2 | 431 | 9053371 | |
| BC150668.1 | - | 2 | 431 | 9087579 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdr2l| Gene Symbol: Cdr2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080929.1 | NP_001074398.1 | 2 | 561 | ||
| GenBank mRNA | BC144823.1 | - | 2 | 431 | 9053371 | |
| BC150668.1 | - | 2 | 431 | 9087579 | ||