siRNA Details
| Gene Symbol: BC068281 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170858.1 | NP_001164329.1 | 2 | 913 | ||
| NM_173416.3 | NP_775592.2 | 2 | 913 | |||
| GenBank mRNA | AK040407.1 | - | 2 | 870 | ||
| AK155130.1 | - | 2 | 833 | |||
| AK164024.1 | - | 3 | 1075 | |||
| AK170427.1 | - | 2 | 811 | |||
| BC068281.1 | - | 2 | 850 | 30008957 | ||

BC068281
Genomic Map