siRNA Details
| Gene Symbol: Arid4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081195.1 | NP_001074664.1 | 8, 9 | 818 | ||
| GenBank mRNA | AK042186.1 | - | 7, 8 | 1007 | ||
| BC025436.1 | - | 8, 9 | 806 | |||
| BC058209.1 | - | 8, 9 | 735 | |||
| BC080780.1 | - | 8, 9 | 753 | |||
| BC157975.1 | - | 7, 8 | 620 | 9007297 | ||
| BC158109.1 | - | 7, 8 | 620 | 9007390 | ||

Arid4a
Genomic Map