siRNA Details
| Gene Symbol: Arid4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081195.1 | NP_001074664.1 | 22, 23 | 3781 | ||
| GenBank mRNA | AK041444.1 | - | 1, 2 | 172 | ||
| BC037715.1 | - | 6, 7 | 1772 | |||
| BC157975.1 | - | 21, 22 | 3580 | 9007297 | ||
| BC158109.1 | - | 21, 22 | 3583 | 9007390 | ||

Arid4a
Genomic Map