siRNA Details
| Gene Symbol: Gpr135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181752.1 | NP_861417.1 | 1 | 456 | ||
| GenBank mRNA | AK132237.1 | - | 591 | |||
| AK141053.1 | - | 1 | 540 | |||
| AY288425.1 | - | 1 | 456 | |||
| BC062104.1 | - | 489 | 6809932 | |||
| BC114990.1 | - | 1 | 455 | |||
| BC115824.1 | - | 1 | 455 | |||
| BC139332.1 | - | 1 | 518 | 8862354 | ||
| BC139335.1 | - | 1 | 518 | 8862357 | ||
| BC148305.2 | - | 1 | 456 | |||
More Information
Other Species Targeted
Set Membership: |
|

Gpr135
Genomic Map