siRNA Details
| Gene Symbol: Bpnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011794.3 | NP_035924.2 | 8, 9 | 896 | ||
| GenBank mRNA | AF125043.1 | - | 8, 9 | 854 | ||
| AK008091.1 | - | 7, 8 | 750 | |||
| AK008714.1 | - | 8, 9 | 877 | |||
| AK093938.1 | - | 8, 9 | 882 | |||
| AK150877.1 | - | 8, 9 | 876 | |||
| AK151931.1 | - | 8, 9 | 900 | |||
| AK152009.1 | - | 8, 9 | 876 | |||
| AK154435.1 | - | 8, 9 | 876 | |||
| BC011036.1 | - | 8, 9 | 849 | 4160742 | ||

Bpnt1
Genomic Map