siRNA Details
| Gene Symbol: Elmsan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163501.1 | NP_001156973.1 | 4 | 2340 | ||
| NM_001163502.1 | NP_001156974.1 | 4 | 2750 | |||
| GenBank mRNA | AK081554.1 | - | 4 | 2764 | ||
| AK155547.1 | - | 4 | 2750 | |||
| AK155768.1 | - | 4 | 2344 | |||
| AK171272.1 | - | 4 | 2755 | |||

Elmsan1
Genomic Map