siRNA Details
| Gene Symbol: Elmsan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163501.1 | NP_001156973.1 | 3 | 1984 | ||
| NM_001163502.1 | NP_001156974.1 | 3 | 2394 | |||
| GenBank mRNA | AK081554.1 | - | 3 | 2408 | ||
| AK155547.1 | - | 3 | 2394 | |||
| AK155768.1 | - | 3 | 1988 | |||
| AK171272.1 | - | 3 | 2397 | |||

Elmsan1
Genomic Map