siRNA Details
| Gene Symbol: Gpr68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177673.1 | NP_001171144.1 | 3 | 1327 | ||
| NM_001177674.1 | NP_001171145.1 | 3 | 1509 | |||
| NM_175493.4 | NP_780702.1 | 3 | 1630 | |||
| GenBank mRNA | AK036659.1 | - | 2 | 1339 | ||
| AK087410.1 | - | 3 | 1311 | |||
| AK154649.1 | - | 2 | 1302 | |||
| AK155886.1 | - | 2 | 1319 | |||
| BC118034.1 | - | 2 | 1308 | 40106885 | ||
| BC118044.1 | - | 2 | 1308 | 40106886 | ||
More Information
Set Membership: |
|

Gpr68
Genomic Map