siRNA Details
| Gene Symbol: Btbd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172806.2 | NP_766394.2 | 8 | 2146 | ||
| GenBank mRNA | BC041669.1 | - | 8 | 2146 | 3482657 | |
| BC057303.1 | - | 7 | 1865 | 5702276 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btbd7| Gene Symbol: Btbd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172806.2 | NP_766394.2 | 8 | 2146 | ||
| GenBank mRNA | BC041669.1 | - | 8 | 2146 | 3482657 | |
| BC057303.1 | - | 7 | 1865 | 5702276 | ||