siRNA Details
| Gene Symbol: Clca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017474.2 | NP_059502.1 | 13 | 2282 | ||
| GenBank mRNA | AB017156.1 | - | 13 | 2265 | ||
| AK007466.1 | - | 13 | 2280 | |||
| AK008659.2 | - | 13 | 2279 | |||
| AK033560.1 | - | 2284 | ||||
| AK136476.1 | - | 13 | 2279 | |||
| BC028343.1 | - | 13 | 2272 | 4217548 | ||
| BC116318.2 | - | 13 | 2265 | 40095991 | ||
| BC116319.2 | - | 13 | 2265 | 40095992 | ||

Clca1
Genomic Map