siRNA Details
Gene Symbol: Pappa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085376.2 | NP_001078845.1 | 2, 3 | 2004 | ||
GenBank mRNA | AK084006.1 | - | 2, 3 | 1515 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pappa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085376.2 | NP_001078845.1 | 2, 3 | 2004 | ||
GenBank mRNA | AK084006.1 | - | 2, 3 | 1515 |