siRNA Details
| Gene Symbol: Def6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027185.3 | NP_081461.2 | 10, 11 | 1727 | ||
| GenBank mRNA | AF329497.1 | - | 10, 11 | 1662 | ||
| AK010356.1 | - | 2, 3 | 201 | |||
| AK038050.1 | - | 10, 11 | 1698 | |||
| AK088460.1 | - | 10, 11 | 1676 | |||
| AY241695.1 | - | 10, 11 | 1662 | |||
| AY278770.1 | - | 4, 5 | 543 | |||
| BC120500.1 | - | 10, 11 | 1727 | 40129423 | ||
| BC131953.1 | - | 10, 11 | 1727 | 40130230 | ||

Def6
Genomic Map