siRNA Details
| Gene Symbol: Dlg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011807.3 | NP_035937.2 | 4, 5 | 541 | ||
| GenBank mRNA | AF388675.1 | - | 4, 5 | 320 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dlg2| Gene Symbol: Dlg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011807.3 | NP_035937.2 | 4, 5 | 541 | ||
| GenBank mRNA | AF388675.1 | - | 4, 5 | 320 | ||