siRNA Details
| Gene Symbol: Zfp58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007575.2 | NP_001007576.1 | 4 | 1105 | ||
| GenBank mRNA | AK041250.1 | - | 4 | 1041 | ||
| AK167399.1 | - | 4 | 1047 | |||
| BC027395.1 | - | 1 | 61 | |||
| BC139150.1 | - | 4 | 971 | 8862172 | ||
| BC139151.1 | - | 4 | 971 | 8862173 | ||

Zfp58
Genomic Map