siRNA Details
| Gene Symbol: Fbln5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011812.4 | NP_035942.1 | 9 | 1253 | ||
| GenBank mRNA | AF112151.1 | - | 9 | 1203 | ||
| AK085170.1 | - | 9 | 1083 | |||
| AK090129.1 | - | 9 | 1192 | |||
| AK159189.1 | - | 9 | 1017 | |||
| AK159471.1 | - | 9 | 1236 | |||
| AK159960.1 | - | 9 | 1223 | |||
| AK165018.1 | - | 9 | 1046 | |||
| AK180395.1 | - | 2 | 54 | |||
| BC006636.1 | - | 9 | 943 | 3482574 | ||

Fbln5
Genomic Map