siRNA Details
| Gene Symbol: Fyb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278269.1 | NP_001265198.1 | 1 | 503 | ||
| NM_011815.5 | NP_035945.1 | 1 | 503 | |||
| GenBank mRNA | AF001863.1 | - | 1 | 479 | ||
| AF061744.1 | - | 1 | 479 | |||
| AK030905.1 | - | 1 | 460 | |||
| AK037454.1 | - | 1 | 504 | |||
| AK038225.1 | - | 1 | 485 | |||
| AK137907.1 | - | 1 | 458 | |||
| AK172013.1 | - | 1 | 459 | |||
| BC064102.1 | - | 1 | 439 | |||
| BC141352.1 | - | 1 | 444 | 9055964 | ||

Fyb
Genomic Map