siRNA Details
| Gene Symbol: Cdc20b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281487.1 | NP_001268416.1 | 6 | 670 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdc20b| Gene Symbol: Cdc20b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281487.1 | NP_001268416.1 | 6 | 670 | ||