siRNA Details
| Gene Symbol: Arhgap22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153800.4 | NP_722495.3 | 10 | 1526 | ||
| GenBank mRNA | AK046097.1 | - | 10 | 1528 | ||
| AK052541.1 | - | 7 | 1172 | |||
| AK078218.1 | - | 9 | 1415 | |||
| AY541447.1 | - | 6 | 1359 | |||
| BC038272.1 | - | 5 | 1024 | 4237331 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK144277.1 | - | 1247 | Chr. - - [-] on Build | ||

Arhgap22
Genomic Map