siRNA Details
| Gene Symbol: Arhgap22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153800.4 | NP_722495.3 | 8, 9 | 1013 | ||
| GenBank mRNA | AK046097.1 | - | 8, 9 | 1015 | ||
| AK052541.1 | - | 5, 6 | 648 | |||
| AK078218.1 | - | 7, 8 | 902 | |||
| AY541447.1 | - | 4, 5 | 846 | |||

Arhgap22
Genomic Map