siRNA Details
| Gene Symbol: Antxrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172808.2 | NP_766396.2 | 12 | 1213 | ||
| GenBank mRNA | AK077206.1 | - | 12 | 1214 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Antxrl| Gene Symbol: Antxrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172808.2 | NP_766396.2 | 12 | 1213 | ||
| GenBank mRNA | AK077206.1 | - | 12 | 1214 | ||