siRNA Details
| Gene Symbol: Lrit2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173418.3 | NP_775594.1 | 3 | 1468 | ||
| GenBank mRNA | AK044383.1 | - | 3 | 1469 | ||
| AK044611.1 | - | 3 | 1470 | |||
| BC037728.1 | - | 3 | 1368 | |||
| BC057628.1 | - | 3 | 1443 | 4511498 | ||

Lrit2
Genomic Map