siRNA Details
Gene Symbol: Cdh24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199470.2 | NP_955764.1 | 6 | 1175 | ||
GenBank mRNA | BC057373.1 | - | 1129 | 6817232 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cdh24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199470.2 | NP_955764.1 | 6 | 1175 | ||
GenBank mRNA | BC057373.1 | - | 1129 | 6817232 |