siRNA Details
| Gene Symbol: Insrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011832.2 | NP_035962.2 | 2 | 716 | ||
| GenBank mRNA | AB007135.1 | - | 2 | 605 | ||
| AK081512.1 | - | 2 | 703 | |||
| AK142578.1 | - | 2 | 717 | |||
| BC137855.1 | - | 2 | 573 | 8860874 | ||

Insrr
Genomic Map