siRNA Details
| Gene Symbol: Sh2b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF086810.1 | - | 1 | 31 | ||
| AF234838.1 | - | 6 | 1419 | |||
| AK017604.1 | - | 6 | 1389 | |||
| AK140319.1 | - | 6 | 1385 | |||
| AK140325.1 | - | 6 | 1385 | |||
| BC049777.1 | - | 6 | 1370 | |||
| BC057334.1 | - | 6 | 1437 | 6811850 | ||

Sh2b2
Genomic Map