siRNA Details
| Gene Symbol: Katna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011835.2 | NP_035965.2 | 9 | 1203 | ||
| GenBank mRNA | AF153197.1 | - | 9 | 1181 | ||
| AK012319.1 | - | 9 | 1202 | |||
| BC009136.1 | - | 9 | 1117 | 3597742 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Katna1| Gene Symbol: Katna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011835.2 | NP_035965.2 | 9 | 1203 | ||
| GenBank mRNA | AF153197.1 | - | 9 | 1181 | ||
| AK012319.1 | - | 9 | 1202 | |||
| BC009136.1 | - | 9 | 1117 | 3597742 | ||