siRNA Details
| Gene Symbol: Katna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011835.2 | NP_035965.2 | 8 | 1130 | ||
| GenBank mRNA | AF153197.1 | - | 8 | 1108 | ||
| AK012319.1 | - | 8 | 1129 | |||
| AK041900.1 | - | 8 | 1125 | |||
| BC009136.1 | - | 8 | 1044 | 3597742 | ||

Katna1
Genomic Map