siRNA Details
| Gene Symbol: Slitrk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175499.4 | NP_780708.1 | 2 | 967 | ||
| GenBank mRNA | AK029275.1 | - | 2 | 890 | ||
| AK044796.1 | - | 2 | 968 | |||
| BC138664.1 | - | 2 | 484 | 8861685 | ||
| BC145927.1 | - | 2 | 484 | 40131230 | ||

Slitrk6
Genomic Map