siRNA Details
| Gene Symbol: Lamc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011836.3 | NP_035966.2 | 21 | 3616 | ||
| GenBank mRNA | AF079520.1 | - | 21 | 3639 | ||
| AF083372.1 | - | 21 | 3553 | |||
| AK132948.1 | - | 18 | 2603 | |||
| BC005535.1 | - | 4 | 361 | |||
| BC096366.1 | - | 21 | 3616 | 2865309 | ||

Lamc3
Genomic Map