siRNA Details
| Gene Symbol: Adamts12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175501.2 | NP_780710.2 | 15 | 2439 | ||
| GenBank mRNA | AJ537452.1 | - | 15 | 2198 | ||
| AK028459.1 | - | 6 | 694 | |||
| AK048612.1 | - | 15 | 2402 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adamts12| Gene Symbol: Adamts12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175501.2 | NP_780710.2 | 15 | 2439 | ||
| GenBank mRNA | AJ537452.1 | - | 15 | 2198 | ||
| AK028459.1 | - | 6 | 694 | |||
| AK048612.1 | - | 15 | 2402 | |||