siRNA Details
| Gene Symbol: Rspo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172815.3 | NP_766403.1 | 4 | 1257 | ||
| GenBank mRNA | AK049891.1 | - | 4 | 1008 | ||
| AK052457.1 | - | 3 | 1381 | |||
| AK087485.1 | - | 4 | 1258 | |||
| BC052844.1 | - | 4 | 608 | |||

Rspo2
Genomic Map