siRNA Details
| Gene Symbol: Mid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011845.2 | NP_035975.1 | 6 | 1187 | ||
| GenBank mRNA | AF196480.1 | - | 6 | 1187 | ||
| AK039479.1 | - | 2 | 795 | |||
| Y18881.1 | - | 7 | 1542 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mid2| Gene Symbol: Mid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011845.2 | NP_035975.1 | 6 | 1187 | ||
| GenBank mRNA | AF196480.1 | - | 6 | 1187 | ||
| AK039479.1 | - | 2 | 795 | |||
| Y18881.1 | - | 7 | 1542 | |||