siRNA Details
| Gene Symbol: Ttc38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033337.4 | NP_001028509.2 | 12, 13 | 1325 | ||
| GenBank mRNA | AK078848.1 | - | 12, 13 | 1287 | ||
| AK133636.1 | - | 11, 12 | 1033 | |||
| BC024550.1 | - | 12, 13 | 1275 | |||
| BC030849.1 | - | 12, 13 | 1288 | |||
| BC132621.1 | - | 12, 13 | 1310 | 40130898 | ||
| BC132623.1 | - | 12, 13 | 1310 | 40130900 | ||

Ttc38
Genomic Map