siRNA Details
| Gene Symbol: Nlrp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF074018.1 | - | 7 | 1638 | ||
| AK087774.1 | - | 7 | 1660 | |||
| AK135731.1 | - | 1 | 599 | |||
| AY329485.1 | - | 7 | 1638 | |||
| AY329486.1 | - | 7 | 1638 | |||
| AY329490.1 | - | 7 | 1638 | |||
| AY329491.1 | - | 7 | 1638 | |||
| BC053384.1 | - | 8 | 1705 | 30049556 | ||

Nlrp5
Genomic Map