siRNA Details
| Gene Symbol: Papss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289477.1 | NP_001276406.1 | 8 | 1020 | ||
| NM_001289478.1 | NP_001276407.1 | 8 | 1207 | |||
| NM_001289479.1 | NP_001276408.1 | 9 | 1318 | |||
| NM_011863.2 | NP_035993.1 | 7 | 1030 | |||
| GenBank mRNA | AK146507.1 | - | 7 | 903 | ||
| AK153296.1 | - | 7 | 900 | |||
| AK160875.1 | - | 9 | 1222 | |||
| AK168982.1 | - | 7 | 903 | |||
| AK169117.1 | - | 8 | 1082 | |||
| AK169124.1 | - | 7 | 897 | |||
| AK169204.1 | - | 8 | 1082 | |||
| BC066055.1 | - | 8 | 1086 | 6833756 | ||
| U34883.1 | - | 7 | 907 | |||

Papss1
Genomic Map