siRNA Details
| Gene Symbol: Papss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289477.1 | NP_001276406.1 | 9 | 1161 | ||
| NM_001289478.1 | NP_001276407.1 | 9 | 1348 | |||
| NM_001289479.1 | NP_001276408.1 | 10 | 1459 | |||
| NM_011863.2 | NP_035993.1 | 8 | 1171 | |||
| GenBank mRNA | AK146507.1 | - | 8 | 1044 | ||
| AK153296.1 | - | 8 | 1041 | |||
| AK160875.1 | - | 10 | 1363 | |||
| AK168982.1 | - | 8 | 1044 | |||
| AK169117.1 | - | 9 | 1223 | |||
| AK169124.1 | - | 8 | 1038 | |||
| AK169204.1 | - | 9 | 1223 | |||
| BC066055.1 | - | 9 | 1227 | 6833756 | ||
| U34883.1 | - | 8 | 1048 | |||

Papss1
Genomic Map