siRNA Details
| Gene Symbol: Mb21d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177718.3 | NP_808386.2 | 2 | 345 | ||
| GenBank mRNA | AK079722.1 | - | 3 | 305 | ||
| BC068245.1 | - | 2 | 333 | |||
| BC085092.1 | - | 2 | 271 | 30735763 | ||
| BE983254.1 | - | 2 | 345 | |||

Mb21d2
Genomic Map