siRNA Details
| Gene Symbol: Zpld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178720.4 | NP_848835.1 | 4 | 468 | ||
| GenBank mRNA | AK034628.1 | - | 4 | 469 | ||
| AK047981.1 | - | 3 | 378 | |||
| AK052599.1 | - | 5 | 578 | |||
| BC116254.1 | - | 4 | 437 | 40093631 | ||
| BC116255.1 | - | 4 | 437 | 40093632 | ||

Zpld1
Genomic Map