siRNA Details
| Gene Symbol: Cadm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145977.1 | NP_001139449.1 | 8, 9 | 1743 | ||
| NM_178721.4 | NP_848836.2 | 9, 10 | 1770 | |||
| GenBank mRNA | AK033973.1 | - | 8, 9 | 1370 | ||
| AK038842.1 | - | 9, 10 | 1749 | |||
| AK043760.1 | - | 9, 10 | 1753 | |||
| AK046800.1 | - | 8, 9 | 1646 | |||
| AK163636.1 | - | 9, 10 | 1747 | |||
| AK165030.1 | - | 9, 10 | 1767 | |||
| BC139401.1 | - | 9, 10 | 1339 | 8862423 | ||
| BC139402.1 | - | 9, 10 | 1339 | 8862424 | ||

Cadm2
Genomic Map