siRNA Details
| Gene Symbol: Cadm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145977.1 | NP_001139449.1 | 3 | 993 | ||
| NM_178721.4 | NP_848836.2 | 4 | 1020 | |||
| GenBank mRNA | AK033973.1 | - | 3 | 620 | ||
| AK038842.1 | - | 4 | 999 | |||
| AK043760.1 | - | 4 | 1003 | |||
| AK046800.1 | - | 3 | 896 | |||
| AK163636.1 | - | 4 | 997 | |||
| AK165030.1 | - | 4 | 1017 | |||
| BC139401.1 | - | 4 | 589 | 8862423 | ||
| BC139402.1 | - | 4 | 589 | 8862424 | ||

Cadm2
Genomic Map